Gene Transcripts

Gene Symbol
Zbtb41
Entrez Gene ID
226470
Gene Name
zinc finger and BTB domain containing 41 homolog
Gene Aliases
8430415N23Rik, 9430031N01, 9830132G07Rik, AI316857
Chromosome Location
Chr. 1: 139421719 - 139453007 on Build GRCm38
UniGene ID
Mm.119711
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172643.5 NP_766231.2 2 1202
GenBank mRNA AK034756.1 - 1 301
AK036544.1 - 2 1201
AK163302.1 - 1 475
BC150859.1 - 1 541 9087770
BC172012.1 - 1 541

View Full Product Details