Gene Transcripts
- Gene Symbol
- Fras1
- Entrez Gene ID
- 231470
- Gene Name
- Fraser syndrome 1 homolog (human)
- Gene Aliases
- E130113P14Rik, bl, mKIAA1500
- Chromosome Location
- Chr. 5: 96373825 - 96784732 on Build GRCm38
- UniGene ID
- Mm.291120
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ489280.1 |
- |
55 |
8771 |
|
AK122518.1 |
- |
8 |
1068 |
|
View Full Product Details