Gene Transcripts

Gene Symbol
Dolk
Entrez Gene ID
227697
Gene Name
dolichol kinase
Gene Aliases
BC026973, Tmem15, mKIAA1094
Chromosome Location
Chr. 2: 30284229 - 30286354 on Build GRCm38
UniGene ID
Mm.26690
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177648.3 NP_808316.1 1 1871
GenBank mRNA AB073968.1 - 1 1814
AK077581.1 - 1 1842
AK144705.1 - 1 1852
AK214790.1 - 1 188
AK217764.1 - 1 146
BC019543.1 - 1 1816
BC026973.1 - 1 1596 5371267

View Full Product Details