Gene Transcripts
- Gene Symbol
- Hmcn2
- Entrez Gene ID
- 665700
- Gene Name
- hemicentin 2
- Gene Aliases
- D030068H01
- Chromosome Location
- Chr. 2: 31313991 - 31460738 on Build GRCm38
- UniGene ID
- Mm.80703
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK083701.1 |
- |
2 |
165 |
|
BC034076.1 |
- |
3 |
595 |
|
BC040753.1 |
- |
3 |
595 |
|
View Full Product Details