Gene Transcripts

Gene Symbol
Coq4
Entrez Gene ID
227683
Gene Name
coenzyme Q4 homolog (yeast)
Gene Aliases
A330041N06, D2Ertd97e
Chromosome Location
Chr. 2: 29788226 - 29797743 on Build GRCm38
UniGene ID
Mm.245948
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178693.4 NP_848808.1 7 1819
GenBank mRNA AK039422.1 - 7 1820
AK163270.1 - 2665

View Full Product Details