Gene Transcripts

Gene Symbol
Radil
Entrez Gene ID
231858
Gene Name
Ras association and DIL domains
Gene Aliases
AI536456, D930005D10Rik
Chromosome Location
Chr. 5: 142484839 - 142551098 on Build GRCm38
UniGene ID
Mm.33766
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289588.1 NP_001276517.1 2 186
NM_178702.4 NP_848817.2 2 186
GenBank mRNA AK038570.1 - 2 119
AK052964.1 - 2 97
AK083410.1 - 2 204
AK154169.1 - 2 125
AK162917.1 - 2 166
AK163852.1 - 2 122
AK173277.1 - 2 150
BC056483.1 - 2 138 5696312
BY201066.1 - 2 188

View Full Product Details