Gene Transcripts

Gene Symbol
Slc6a1
Entrez Gene ID
232333
Gene Name
solute carrier family 6 (neurotransmitter transporter, GABA), member 1
Gene Aliases
A730043E01, GABATHG, GABATR, GAT-1, Gabt, Gabt1, Gat1, XT-1, Xtrp1
Chromosome Location
Chr. 6: 114282635 - 114317525 on Build GRCm38
UniGene ID
Mm.5260
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178703.4 NP_848818.1 8, 9 1221
GenBank mRNA AK042956.1 - 8, 9 1083
AK047663.1 - 8, 9 1067
AK052971.1 - 8, 9 1046
AK053883.1 - 8, 9 1043
BC059080.1 - 8, 9 1072 6827121
M92378.1 - 7 980

View Full Product Details