Gene Transcripts

Gene Symbol
Iqsec1
Entrez Gene ID
232227
Gene Name
IQ motif and Sec7 domain 1
Gene Aliases
AW561907, BC026481, BRAG2, D6Ertd349e, cI-43
Chromosome Location
Chr. 6: 90659596 - 90988460 on Build GRCm38
UniGene ID
Mm.196943
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134383.1 NP_001127855.1 14 3808
NM_001134384.1 NP_001127856.1 14 5772
GenBank mRNA AK044711.1 - 1 1974

View Full Product Details