Gene Transcripts

Gene Symbol
Paqr3
Entrez Gene ID
231474
Gene Name
progestin and adipoQ receptor family member III
Gene Aliases
6330415A20Rik, AY424292, RKTG
Chromosome Location
Chr. 5: 97082329 - 97111748 on Build GRCm38
UniGene ID
Mm.332505
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198422.2 NP_940814.1 5, 6 954
GenBank mRNA AK149213.1 - 5, 6 933
AK172689.1 - 5, 6 910
AY424292.1 - 5, 6 954
BC120853.1 - 5, 6 939 40129776
BC120855.1 - 5, 6 940 40129778

View Full Product Details