Gene Transcripts

Gene Symbol
U2af1
Entrez Gene ID
108121
Gene Name
U2 small nuclear ribonucleoprotein auxiliary factor (U2AF) 1
Gene Aliases
2010107D16Rik, 35kDa
Chromosome Location
Chr. 17: 31647081 - 31658764 on Build GRCm38
UniGene ID
Mm.379289
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163769.1 NP_001157241.1 6 442
NM_024187.4 NP_077149.2 6 442
GenBank mRNA AK008332.1 - 6 404
AK012334.1 - 6 402
AK012849.1 - 6 402
AK088979.1 - 6 412
AK150872.1 - 6 442
AV453783.1 - 6 424
BC002184.1 - 6 381
BC030413.1 - 3 1243
BC033478.1 - 4 210
BC069888.1 - 6 384
BC115479.1 - 6 404 40100558
BC115480.1 - 6 404 40100559

View Full Product Details