Gene Transcripts

Gene Symbol
Pwp2
Entrez Gene ID
110816
Gene Name
PWP2 periodic tryptophan protein homolog (yeast)
Gene Aliases
6530411D08Rik, Pwp2h, wdp103
Chromosome Location
Chr. 10: 78170910 - 78185171 on Build GRCm38
UniGene ID
Mm.103522
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029546.2 NP_083822.1 5 414
GenBank mRNA AB041855.1 - 5 375
AK088254.1 - 5 415
AK088983.1 - 5 415
BC112325.1 - 5 380 5355810
BC117903.1 - 5 395
BC128480.1 - 5 395 40097069

View Full Product Details