Gene Transcripts
- Gene Symbol
- Foxp1
- Entrez Gene ID
- 108655
- Gene Name
- forkhead box P1
- Gene Aliases
- 3110052D19Rik, 4932443N09Rik, AI461938, AW494214
- Chromosome Location
- Chr. 6: 98925342 - 99523024 on Build GRCm38
- UniGene ID
- Mm.234965
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001197321.1 |
NP_001184250.1
|
9, 10 |
1215 |
|
NM_001197322.1 |
NP_001184251.1
|
6, 7 |
822 |
|
NM_053202.2 |
NP_444432.1
|
10, 11 |
1367 |
|
GenBank mRNA |
AB221601.1 |
- |
7, 8 |
1062 |
|
AB221602.1 |
- |
7, 8 |
1060 |
|
AB594192.1 |
- |
11, 12 |
1775 |
|
AF339103.1 |
- |
7, 8 |
1056 |
|
AF339104.1 |
- |
7, 8 |
1056 |
|
AF339105.1 |
- |
|
836 |
|
AK031837.1 |
- |
3, 4 |
359 |
|
AK077062.1 |
- |
6, 7 |
717 |
|
AK139831.1 |
- |
6, 7 |
709 |
|
BC064764.1 |
- |
9, 10 |
1201 |
30430449 |
EF410101.1 |
- |
1, 2 |
357 |
|
View Full Product Details