Gene Transcripts

Gene Symbol
Egln1
Entrez Gene ID
112405
Gene Name
egl-9 family hypoxia-inducible factor 1
Gene Aliases
AI503754, C1orf12, HIF-PH2, HPH-2, Hif-p4h-2, ORF13, Phd2, SM-20
Chromosome Location
Chr. 8: 124908586 - 124949319 on Build GRCm38
UniGene ID
Mm.140619
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053207.2 NP_444437.2 3, 4 1287
GenBank mRNA AF453878.1 - 3, 4 476
AJ310546.1 - 3, 4 990
BC006903.1 - 3, 4 287 3600175

View Full Product Details