Gene Transcripts

Gene Symbol
Muc4
Entrez Gene ID
140474
Gene Name
mucin 4
Gene Aliases
4933405I11Rik, Asgp
Chromosome Location
Chr. 16: 32735886 - 32782391 on Build GRCm38
UniGene ID
Mm.214599
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080457.3 NP_536705.3 2 624
GenBank mRNA AF441786.1 - 235
AY007202.1 - 2 627

View Full Product Details