Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167991.1 | NP_001161463.1 | 18 | 1796 | |
NM_133255.2 | NP_573518.2 | 18 | 1796 | ||
GenBank mRNA | AK030599.1 | - | 20 | 1812 | |
AK042048.1 | - | 18 | 1784 | ||
AK136069.1 | - | 19 | 1718 | ||
AK145768.1 | - | 18 | 1764 | ||
AK166954.1 | - | 18 | 1776 | ||
AK169073.1 | - | 18 | 1764 | ||
BC002226.1 | - | 10 | 1005 | ||
BC019486.1 | - | 18 | 1772 | 4211990 | |
BC026609.1 | - | 10 | 956 | ||
BC029967.1 | - | 18 | 1737 | ||
BC049908.1 | - | 1 | 333 | ||
BC055881.1 | - | 18 | 1742 | 3590336 | |
BC080744.1 | - | 18 | 1786 | 30471369 |