Gene Transcripts

Gene Symbol
Rnf44
Entrez Gene ID
105239
Gene Name
ring finger protein 44
Gene Aliases
AI854545, mKIAA1100
Chromosome Location
Chr. 13: 54679399 - 54693995 on Build GRCm38
UniGene ID
Mm.25366
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146025.1 NP_001139497.1 7 1259
NM_001146026.1 NP_001139498.1 7 1184
NM_001146027.1 NP_001139499.1 7 1050
NM_134064.2 NP_598825.2 7 1192
GenBank mRNA AK147349.1 - 7 1185
AK160090.1 - 7 1043
AK160115.1 - 7 1043
AK161651.1 - 7 1172
AK171943.1 - 7 1041
BC017630.1 - 7 985 4507289
BC035548.1 - 7 1007 4221284

View Full Product Details