Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005908.3 | NP_005899.3 | 6 | 926 | |
XM_017008203.1 | XP_016863692.1 | 7 | 912 | ||
XM_017008204.1 | XP_016863693.1 | 5 | 757 | ||
GenBank mRNA | AB209846.1 | - | 7 | 951 | |
AK291598.1 | - | 6 | 914 | ||
AK300015.1 | - | 5 | 696 | ||
AK312827.1 | - | 6 | 881 | ||
BC015743.1 | - | 6 | 916 | 4905535 | |
EU009130.1 | - | 6 | 826 | ||
U60337.1 | - | 6 | 913 |