Gene Transcripts

Gene Symbol
Kdelr3
Entrez Gene ID
105785
Gene Name
KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 3
Gene Aliases
AI173274
Chromosome Location
Chr. 15: 79516408 - 79527739 on Build GRCm38
UniGene ID
Mm.298199
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134090.2 NP_598851.2 5 819
GenBank mRNA BC011472.1 - 5 759
BC024420.1 - 5 752 4985334

View Full Product Details