Gene Transcripts

Gene Symbol
Fam50a
Entrez Gene ID
108160
Gene Name
family with sequence similarity 50, member A
Gene Aliases
D0HXS9928E, XAP-5
Chromosome Location
Chr. X: 74313033 - 74320149 on Build GRCm38
UniGene ID
Mm.4370
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138607.3 NP_613073.2 7 795
GenBank mRNA AK008402.1 - 7 796
BC026128.1 - 6 1168
BC125297.1 - 6 661 40129812
Y18505.1 - 7 760

View Full Product Details