Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138952.3 | NP_620402.1 | 2 | 371 | |
GenBank mRNA | AF461040.1 | - | 2 | 269 | |
AF487539.1 | - | 2 | 269 | ||
AK159593.1 | - | 2 | 395 | ||
AK172319.1 | - | 2 | 393 | ||
AK172417.1 | - | 2 | 393 | ||
BC050845.1 | - | 2 | 384 | ||
BC058088.1 | - | 2 | 516 | ||
BC069878.1 | - | 2 | 396 | 5316066 |