Gene Transcripts

Gene Symbol
Gps1
Entrez Gene ID
209318
Gene Name
G protein pathway suppressor 1
Gene Aliases
Cops1, Csn1, R75577, Sgn1
Chromosome Location
Chr. 11: 120784260 - 120789107 on Build GRCm38
UniGene ID
Mm.30195
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177874.1 NP_001171345.1 3 220
NM_145370.2 NP_663345.2 3 360
GenBank mRNA AK088490.1 - 3 217
AK190400.1 - 2 103
AK204171.1 - 2 104
BC003350.1 - 3 360 3481979
BC103782.1 - 3 187 6486599

View Full Product Details