Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172157.1 | NP_001165628.1 | 3, 4 | 382 | |
NM_153530.2 | NP_705758.1 | 3, 4 | 382 | ||
GenBank mRNA | AK029652.1 | - | 3, 4 | 359 | |
AK031180.1 | - | 3, 4 | 365 | ||
AK040613.1 | - | 3, 4 | 359 | ||
BC036177.1 | - | 3, 4 | 343 | 5004341 | |
CJ063044.1 | - | 3, 4 | 361 |