Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161767.1 | NP_001155239.1 | 11 | 2093 | |
NM_001161768.1 | NP_001155240.1 | 11 | 2090 | ||
NM_172451.3 | NP_766039.2 | 11 | 2089 | ||
GenBank mRNA | AJ133523.1 | - | 9 | 1705 | |
AK028506.1 | - | 11 | 2089 | ||
AK049222.1 | - | 11 | 2092 | ||
AK155008.1 | - | 11 | 2096 | ||
AK159721.1 | - | 11 | 2071 | ||
BC119324.1 | - | 9 | 1742 | 8734073 | |
BC119326.1 | - | 9 | 1742 | 8734075 |