Gene Transcripts

Gene Symbol
Prr9
Entrez Gene ID
109314
Gene Name
proline rich 9
Gene Aliases
A030004J04Rik
Chromosome Location
Chr. 3: 92122204 - 92123947 on Build GRCm38
UniGene ID
Mm.440617
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175424.3 NP_780633.1 2 336
GenBank mRNA AK079463.1 - 2 337

View Full Product Details