Gene Transcripts

Gene Symbol
Kctd12b
Entrez Gene ID
207474
Gene Name
potassium channel tetramerisation domain containing 12b
Gene Aliases
9530071H11Rik, B430319H24Rik
Chromosome Location
Chr. X: 153685154 - 153696386 on Build GRCm38
UniGene ID
Mm.271572
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175429.3 NP_780638.1 3 2190
GenBank mRNA AK046708.1 - 3 2118
AK085035.1 - 3 2122
BC064459.1 - 3 2190 6821736

View Full Product Details