Gene Transcripts

Gene Symbol
Slc30a9
Entrez Gene ID
109108
Gene Name
solute carrier family 30 (zinc transporter), member 9
Gene Aliases
2310024J23Rik, AL024256, GAC63, HUEL
Chromosome Location
Chr. 5: 67306937 - 67356834 on Build GRCm38
UniGene ID
Mm.234455
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178651.3 NP_848766.2 5 620
GenBank mRNA AF263460.1 - 3 174
AK028885.1 - 5 587
AK035541.1 - 5 607
AK038559.1 - 4 526
AK082853.1 - 5 622
AK143277.1 - 5 621
AY682914.1 - 6 613
BC026565.1 - 1 38
BC027806.1 - 5 382
BC031705.1 - 2 134
BC055773.1 - 2 73
BC078440.1 - 3 197
BC141008.1 - 6 586 9055620

View Full Product Details