Gene Transcripts

Gene Symbol
Tnfrsf22
Entrez Gene ID
79202
Gene Name
tumor necrosis factor receptor superfamily, member 22
Gene Aliases
2810028K06Rik, C130035G06Rik, SOBa, Tnfrh2, Tnfrsf1al2, mDcTrailr2
Chromosome Location
Chr. 7: 143634808 - 143649752 on Build GRCm38
UniGene ID
Mm.261384
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023680.3 NP_076169.2 7 1691
GenBank mRNA AK012838.1 - 7 1692

View Full Product Details