Gene Transcripts

Gene Symbol
Elp4
Entrez Gene ID
77766
Gene Name
elongation protein 4 homolog (S. cerevisiae)
Gene Aliases
A330107A17Rik, Paxneb
Chromosome Location
Chr. 2: 105700109 - 105904500 on Build GRCm38
UniGene ID
Mm.234998
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023876.3 NP_076365.2 4 473
GenBank mRNA AJ276004.1 - 4 472
AK020742.1 - 4 481
AK034040.1 - 4 478
AK034529.1 - 4 479
AK037104.1 - 4 497
AK039447.1 - 4 485
AK039758.1 - 4 474
AK079069.1 - 4 488
AK168964.1 - 4 537
EF410086.1 - 337

View Full Product Details