Gene Transcripts

Gene Symbol
Popdc3
Entrez Gene ID
78977
Gene Name
popeye domain containing 3
Gene Aliases
AA682104, Pop3
Chromosome Location
Chr. 10: 45289305 - 45318450 on Build GRCm38
UniGene ID
Mm.24748
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024286.1 NP_077248.1 4 1672
GenBank mRNA AK052325.1 - 4 1672

View Full Product Details