Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256100.1 | NP_001243029.1 | 3 | 622 | |
NM_001256101.1 | NP_001243030.1 | 3 | 611 | ||
NM_001256102.1 | NP_001243031.1 | 2 | 451 | ||
NM_029581.4 | NP_083857.1 | 2 | 462 | ||
GenBank mRNA | AK012733.1 | - | 3 | 589 | |
AK147090.1 | - | 2 | 440 | ||
BC052439.1 | - | 2 | 416 | 6406303 | |
BC115684.1 | - | 2 | 403 | 40104230 | |
BC115685.1 | - | 3 | 574 | 40104232 |