Gene Transcripts

Gene Symbol
Hnrnpm
Entrez Gene ID
76936
Gene Name
heterogeneous nuclear ribonucleoprotein M
Gene Aliases
2610023M21Rik, AA409009, Hnrpm, mKIAA4193
Chromosome Location
Chr. 17: 33646233 - 33685458 on Build GRCm38
UniGene ID
Mm.311439
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029804.3 NP_084080.1 6 550
GenBank mRNA AK011521.1 - 6 496
AK048220.1 - 1 1131
AK166670.1 - 6 485
BC054785.1 - 1952

View Full Product Details