Gene Transcripts
- Gene Symbol
- Efcab6
- Entrez Gene ID
- 77627
- Gene Name
- EF-hand calcium binding domain 6
- Gene Aliases
- 4931407K02Rik, 4932408N08Rik, bM150J22.2, mKIAA1672
- Chromosome Location
- Chr. 15: 83866712 - 84075025 on Build GRCm38
- UniGene ID
- Mm.196288
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029946.4 |
NP_084222.4
|
10 |
1208 |
|
GenBank mRNA |
BC065111.1 |
- |
10 |
1208 |
|
View Full Product Details