Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030215.3 | NP_084491.3 | 6 | 1891 | |
GenBank mRNA | AB056151.1 | - | 6 | 1740 | |
AF208046.1 | - | 6 | 1136 | ||
AK041886.1 | - | 6 | 1847 | ||
AK050368.1 | - | 6 | 1893 | ||
AK082078.1 | - | 5 | 2469 | ||
AK167570.1 | - | 6 | 1892 | ||
AK170542.1 | - | 6 | 1884 | ||
AK170846.1 | - | 6 | 1872 | ||
AK205588.1 | - | 2 | 129 | ||
AK209893.1 | - | 3 | 240 | ||
BC010482.1 | - | 6 | 1883 | 3492125 | |
BC058744.1 | - | 6 | 1281 |