Gene Transcripts
- Gene Symbol
- Dlst
- Entrez Gene ID
- 78920
- Gene Name
- dihydrolipoamide S-succinyltransferase (E2 component of 2-oxo-glutarate complex)
- Gene Aliases
- 1600017E01Rik, 4632413C10Rik, 4930529O08Rik, DLTS
- Chromosome Location
- Chr. 12: 85110824 - 85134091 on Build GRCm38
- UniGene ID
- Mm.296221
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030225.4 |
NP_084501.1
|
10, 11 |
834 |
|
GenBank mRNA |
AK019713.2 |
- |
|
831 |
|
AK054053.1 |
- |
10, 11 |
788 |
|
AK149664.1 |
- |
10, 11 |
810 |
|
AK158877.1 |
- |
10, 11 |
835 |
|
AK168570.1 |
- |
10, 11 |
810 |
|
AK169943.1 |
- |
10, 11 |
788 |
|
AK197945.1 |
- |
1, 2 |
37 |
|
AK203473.1 |
- |
1, 2 |
37 |
|
AK205241.1 |
- |
1, 2 |
37 |
|
AK214016.1 |
- |
1, 2 |
91 |
|
AK220022.1 |
- |
1, 2 |
37 |
|
BC006702.1 |
- |
10, 11 |
771 |
3583936 |
CT010197.1 |
- |
10, 11 |
770 |
|
View Full Product Details