Gene Transcripts

Gene Symbol
Dlst
Entrez Gene ID
78920
Gene Name
dihydrolipoamide S-succinyltransferase (E2 component of 2-oxo-glutarate complex)
Gene Aliases
1600017E01Rik, 4632413C10Rik, 4930529O08Rik, DLTS
Chromosome Location
Chr. 12: 85110824 - 85134091 on Build GRCm38
UniGene ID
Mm.296221
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030225.4 NP_084501.1 15 2511
GenBank mRNA AK149664.1 - 15 2487
AK158877.1 - 15 2512
AK168570.1 - 15 2487
AK169943.1 - 15 2465
AK201762.1 - 1 109
AK202210.1 - 1 109
AK205589.1 - 1 110
AK206354.1 - 1 109
AK207941.1 - 1 109
AK209363.1 - 1 109
AK216542.1 - 1 383
AK219425.1 - 1 123
BC006702.1 - 15 2448 3583936
BC024066.1 - 6 1898
CJ308553.1 - 1 160

View Full Product Details