Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030559.3 | NP_085036.3 | 13 | 1333 | |
GenBank mRNA | AB056721.1 | - | 13 | 1291 | |
AK032558.1 | - | 4 | 294 | ||
AK049872.1 | - | 13 | 1290 | ||
AK162984.1 | - | 11 | 1529 | ||
AK169694.1 | - | 10 | 1870 | ||
AK177879.1 | - | 2 | 157 | ||
BC003944.1 | - | 1 | 205 | 3487257 | |
BC025626.1 | - | 13 | 1299 | 5320572 |