Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030714.2 | NP_109639.1 | 6 | 1728 | |
GenBank mRNA | AB015425.1 | - | 5 | 1711 | |
AK205963.1 | - | 1 | 301 | ||
AK212525.1 | - | 1 | 314 | ||
AK213413.1 | - | 1 | 314 | ||
AK217286.1 | - | 1 | 272 | ||
BC048434.1 | - | 3 | 238 | ||
BC099687.1 | - | 6 | 1728 | 6311735 | |
BC138304.1 | - | 5 | 1608 | 8861323 | |
BC157899.1 | - | 5 | 1610 | 9088091 |