Gene Transcripts

Gene Symbol
Tmem2
Entrez Gene ID
83921
Gene Name
transmembrane protein 2
Gene Aliases
Chromosome Location
Chr. 19: 21777508 - 21858437 on Build GRCm38
UniGene ID
Mm.329776
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033759.2 NP_001028931.1 7 1898
NM_031997.4 NP_114386.3 8 1980
GenBank mRNA AK129352.1 - 2 199
BC076570.1 - 7 1504 6830791
BC089353.1 - 7 1880 6410437
BQ895808.1 - 200

View Full Product Details