Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032396.3 | NP_115772.2 | 4 | 497 | |
GenBank mRNA | AB059617.1 | - | 4 | 460 | |
AK083565.1 | - | 2 | 105 | ||
AK141321.1 | - | 4 | 509 | ||
BC049771.1 | - | 4 | 421 | ||
BC082546.1 | - | 4 | 497 | 6855459 | |
BC138464.1 | - | 4 | 505 | 8861484 | |
BC138465.1 | - | 4 | 505 | 8861485 |