Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033595.4 | NP_291073.1 | 1 | 1787 | |
GenBank mRNA | AY013795.1 | - | 1 | 1250 | |
BC054528.1 | - | 1 | 1378 | 5719201 | |
BC056998.1 | - | 1 | 1787 | 6831906 | |
BC066851.1 | - | 1 | 1594 | 30086762 | |
U88550.1 | - | 1 | 1343 |