Gene Transcripts

Gene Symbol
Igsf9
Entrez Gene ID
93842
Gene Name
immunoglobulin superfamily, member 9
Gene Aliases
644ETD8, Dasm1, Kiaa1355-hp, NRT1, Ncaml, mKIAA1355
Chromosome Location
Chr. 1: 172481776 - 172498877 on Build GRCm38
UniGene ID
Mm.214530
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145800.1 NP_001139272.1 12 1751
NM_033608.3 NP_291086.2 12 1817
GenBank mRNA AF317839.1 - 12 1817
AK078536.1 - 12 1745
AK145121.1 - 12 1726
BC034594.1 - 12 1733
BC040281.1 - 12 1777 5364073

View Full Product Details