Gene Transcripts

Gene Symbol
Asb15
Entrez Gene ID
78910
Gene Name
ankyrin repeat and SOCS box-containing 15
Gene Aliases
4930400E23Rik, Asb-15
Chromosome Location
Chr. 6: 24528144 - 24573164 on Build GRCm38
UniGene ID
Mm.160150
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080847.3 NP_543123.2 2 215
GenBank mRNA AF403043.1 - 2 216
AK019556.1 - 2 216
AK029759.1 - 2 147
AK079418.1 - 2 321
BC050794.1 - 2 164 6742480

View Full Product Details