Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133351.3 | NP_579929.1 | 4 | 551 | |
GenBank mRNA | AB038244.1 | - | 4 | 532 | |
AF188613.1 | - | 4 | 465 | ||
AF378085.2 | - | 4 | 542 | ||
AK010640.1 | - | 4 | 434 | ||
AK078696.1 | - | 4 | 506 | ||
AY335911.1 | - | 5 | 399 | ||
BC003851.1 | - | 4 | 525 | 3600399 | |
CB600036.1 | - | 4 | 554 |