Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133798.3 | NP_598559.2 | 9 | 2249 | |
GenBank mRNA | AK138497.1 | - | 1525 | ||
AK146901.1 | - | 9 | 2130 | ||
AK147922.1 | - | 9 | 2110 | ||
AK158201.1 | - | 9 | 2140 | ||
AK162206.1 | - | 8 | 2017 | ||
AK164329.1 | - | 9 | 2125 | ||
AK167692.1 | - | 9 | 2250 | ||
BC018508.1 | - | 8 | 1792 | 3156447 |