Gene Transcripts

Gene Symbol
Plekha1
Entrez Gene ID
101476
Gene Name
pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 1
Gene Aliases
AA960558, C920009D07Rik, TAPP1
Chromosome Location
Chr. 7: 130865729 - 130913308 on Build GRCm38
UniGene ID
Mm.323554
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133942.2 NP_598703.1 10 3160
GenBank mRNA BC020017.1 - 10 3155
BC024470.1 - 1 1304

View Full Product Details