Gene Transcripts

Gene Symbol
Numa1
Entrez Gene ID
101706
Gene Name
nuclear mitotic apparatus protein 1
Gene Aliases
6720401E04Rik, AA764025, AL022610, AU014979
Chromosome Location
Chr. 7: 101934002 - 102014959 on Build GRCm38
UniGene ID
Mm.27259
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133947.3 NP_598708.3 24, 25 6431
GenBank mRNA AK144141.1 - 12, 13 2218
BC004667.1 - 5, 6 700
BC006631.1 - 12, 13 2204
BC049791.1 - 24, 25 6431 6505037

View Full Product Details