Gene Transcripts
- Gene Symbol
- Tmem40
- Entrez Gene ID
- 94346
- Gene Name
- transmembrane protein 40
- Gene Aliases
- 9030407H20Rik, AV091463, W64236
- Chromosome Location
- Chr. 6: 115729131 - 115762466 on Build GRCm38
- UniGene ID
- Mm.29739
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001168256.1 |
NP_001161728.1
|
11, 12 |
868 |
|
NM_001168257.1 |
NP_001161729.1
|
10, 11 |
793 |
|
NM_001168258.1 |
NP_001161730.1
|
6, 7 |
593 |
|
NM_001168259.1 |
NP_001161731.1
|
7, 8 |
360 |
|
NM_144805.2 |
NP_659054.2
|
11, 12 |
736 |
|
GenBank mRNA |
AK078864.1 |
- |
7, 8 |
361 |
|
BC019416.1 |
- |
11, 12 |
698 |
5151159 |
CT010303.1 |
- |
10, 11 |
666 |
|
CV561368.1 |
- |
10, 11 |
735 |
|
CX564997.1 |
- |
11, 12 |
767 |
|
View Full Product Details