Gene Transcripts

Gene Symbol
Zdhhc16
Entrez Gene ID
74168
Gene Name
zinc finger, DHHC domain containing 16
Gene Aliases
1500015N03Rik, Aph2
Chromosome Location
Chr. 19: 41933472 - 41944104 on Build GRCm38
UniGene ID
Mm.20387
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023740.2 NP_076229.2 8 1087
GenBank mRNA AF176814.1 - 8 1060
AK005255.1 - 7 819
AK151505.1 - 8 1069
AK168042.1 - 8 1062
AK181099.1 - 1 51
AK193583.1 - 1 51
AK206660.1 - 1 51
AK219891.1 - 1 51
BC010835.1 - 8 1051 4239913

View Full Product Details