Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320049.1 | NP_001306978.1 | 4, 5 | 622 | |
NM_006331.7 | NP_006322.4 | 5, 6 | 772 | ||
GenBank mRNA | AK291597.1 | - | 5, 6 | 637 | |
BC055314.1 | - | 5, 6 | 664 | 4718702 | |
BP315329.1 | - | 4, 5 | 490 | ||
U72514.1 | - | 5, 6 | 615 |