Gene Transcripts

Gene Symbol
Mro
Entrez Gene ID
71263
Gene Name
maestro
Gene Aliases
4930507C04Rik, 4933435E20Rik
Chromosome Location
Chr. 18: 73851827 - 73881334 on Build GRCm38
UniGene ID
Mm.126220
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK017067.1 - 6 1469
AK140653.1 - 1 666
AY243876.1 - 9 1861

View Full Product Details